Pesticide standard, Propiconazole, reference material, Honeywell Fluka™
Catalog # HONCR1617-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:0,1 mL
- Standards:Analytical Standard
- CAS No.:60207-90-1
- Description:Propiconazole
- Formula:C₁₅H₁₇Cl₂N₃O₂
- Storage temperature:2-8°C
- Description:Propiconazole
- Form:Viscous liquid
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.