Order Entry
Denmark
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Non-volatile Acid Standard Mix
  46985-U
 :  Merck
 :  
Supelco® Organic Reference Standard, Non-volatile Acid Standard Mix
  46985-U
 :  Merck
 :  

 

  • Concentration:
    0,01 meq/ml each component in deionised water
  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    Non-volatile acid standard mix
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100