Order Entry
Denmark
ContactUsLinkComponent
 
EPA 8270 Phenols mix, certified reference material, Supelco®
Catalog # 47377.
Supplier:  Merck
CAS Number:  
EPA 8270 Phenols mix, certified reference material, Supelco®
Catalog # 47377.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 mug/mL each component in isopropanol
  • Pk:
    1 mL
  • Description:
    EPA 8270 Phenols Mix
  • Type:
    Certified reference material

Specifications

About this item