Supelco® PESTANAL® Organic Reference Standard, Triadimenol-(4-chlorophenoxy-d₄)
Catalog # 89947-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:PESTANAL®, analytical standard
- Description:Triadimenol-(4-chlorophenoxy-d₄)
- Storage conditions:2…8 °C
- Hazard class description:H302 + H332 - H412
- Shipping conditions:STD
- MDL No.:MFCD27992127
- M-Clarity quality level:MQ100
- Synonym(s):α-tert-Butyl-β-(4-chlorophenoxy-d₄)-1H-1,2,4-triazole-1-ethanol; Triadimenol-d₄
- Molecular weight:299,79
Specifications
About this item
- GHS signal: GHS07
Caution: P273 - P301 + P312 + P330 - P304 + P340 + P312