Supelco® PESTANAL® Organic Reference Standard, Propetamphos
34371-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:31218-83-4
- Description:Propetamphos
- Storage conditions:2...8 °C
- Hazard class description:H301 + H311 - H330 - H410
- Shipping conditions:STD
- MDL No.:MFCD00137384
- M-Clarity quality level:MQ100
- Formula:C₁₀H₂₀NO₄PS
- Molecular weight:281,31
- Storage temperature:2...8 °C
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: P260 - P264 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310