Supelco® Organic Reference Standard, PCB No 52 Solution
31297-2ML
: Merck
:
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:35693-99-3
- Description:PCB No 52 solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055544
- M-Clarity quality level:MQ100
- Synonym(s):2,2′,5,5′-Tetrachlorobiphenyl solution; 2,2′,5,5′-PCB
- Formula:C₁₂H₆Cl₄
- Molecular weight:291,99
- Storage temperature:2...8 °C
- Flash point:–12 °C - closed cup
- GHS signal: GHS02, GHS08, GHS07, GHS09
: P210 - P273 - P301 + P310 - P303 + P361 + P353 - P314 - P331