Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, N,N-Dimethylallylamine, analytical standard, Supelco®
Organic reference standard, N,N-Dimethylallylamine, analytical standard, Supelco®
Catalog # 01987-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, N,N-Dimethylallylamine, analytical standard, Supelco®
Catalog # 01987-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    2155-94-4
  • Description:
    N,N-Dimethylallylamine
  • Formula:
    C₅H₁₁N
  • Description:
    N,N-Dimethylallylamine

Specifications

About this item