Order Entry
Denmark
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Metaflumizone
Supelco® PESTANAL® Organic Reference Standard, Metaflumizone
Catalog # 32966-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Metaflumizone
Catalog # 32966-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    139968-49-3
  • Description:
    Metaflumizone
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H361fd - H362 - H373
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD08690508
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2′-[2-(4-Cyanophenyl)-1-(α,α,α-trifluoro-m-tolyl)ethylidene]-4-(trifluoromethoxy)carbanilohydrazide
  • Formula:
    C₂₄H₁₆F₆N₄O₂
  • Molecular weight:
    506,40
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS09
Caution: P202 - P260 - P263 - P264 - P270 - P308 + P313