Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Hexamethylcyclotrisiloxane, analytical standard, Supelco®
Organic reference standard, Hexamethylcyclotrisiloxane, analytical standard, Supelco®
Catalog # 38677-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Hexamethylcyclotrisiloxane, analytical standard, Supelco®
Catalog # 38677-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    541-05-9
  • Description:
    Hexamethylcyclotrisiloxane
  • Melting point:
    50...64 °C (lit.)
  • Formula:
    C₆H₁₈O₃Si₃
  • Description:
    Hexamethylcyclotrisiloxane
  • Boiling point:
    134 °C(lit.)

Specifications

About this item