Supelco® PESTANAL® Organic Reference Standard, Glyphosate
45521-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1071-83-6
- Description:Glyphosate
- Storage conditions:2...30 °C
- Hazard class description:H312 - H318 - H411
- Shipping conditions:STD
- MDL No.:MFCD00055350
- M-Clarity quality level:MQ100
- Synonym(s):N-(Phosphonomethyl)glycine; Glyphosate
- Melting point:230 °C (dec.) (lit.)
- Formula:(HO)₂P(O)CH₂NHCH₂CO₂H
- Molecular weight:169,07
- Flash point:Not applicable
- GHS signal: GHS05, GHS09
: P273 - P280 - P302 + P352 + P312 - P305 + P351 + P338 - P362 + P364 - P391