Supelco® Organic Reference Standard, Erucamide
Erucamide, Analytical standard, 25 mg
Catalog # 90082-25MG
Supplier: Merck
Jump to:
Product Details & Documents
- GHS signal: GHS07
Specifications
Specifications
- Pk:25 mg
- Standards:Analytical standard
- Description:Erucamide
- Cat. No.:90082-25MG
- M-Clarity quality level:MQ100
- Formula:CH₃(CH₂)₇CH=CH(CH₂)₁₁CONH₂
- Storage conditions:–20 °C
- MDL No.:MFCD00882379
- Melting point:75 - 85 °C; 79 - 81 °C (lit.)
- Synonym(s):cis-13-Docosenoamide; Erucamide
- Molecular weight:337,58
- CAS No.:112-84-5
- Shipping conditions:WICE



