Order Entry
Denmark
ContactUsLinkComponent
Pesticide reference standard, Cylindrospermopsin solution, analytical standard, Supelco®
Pesticide reference standard, Cylindrospermopsin solution, analytical standard, Supelco®
Catalog # 32087-1ML
Supplier:  Merck
CAS Number:  
undefined
Pesticide reference standard, Cylindrospermopsin solution, analytical standard, Supelco®
Catalog # 32087-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in H2O
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    143545-90-8
  • Description:
    Cylindrospermopsin solution
  • Formula:
    C₁₅H₂₁N₅O₇S
  • Storage temperature:
    −20 °C
  • Description:
    Cylindrospermopsin solution

Specifications

About this item