Organic reference standard, clofibrate, analytical standard, Supelco®
Catalog # 41326-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:637-07-0
- Description:Clofibrate
- Formula:C₁₂H₁₅ClO₃
- Description:Clofibrate