Order Entry
Denmark
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Climbazole
Supelco® PESTANAL® Organic Reference Standard, Climbazole
Catalog # 36127-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Climbazole
Catalog # 36127-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    38083-17-9
  • Description:
    Climbazole
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055505
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethylbutan-2-one
  • Melting point:
    96...100 °C
  • Formula:
    C₁₅H₁₇ClN₂O₂
  • Molecular weight:
    292,76
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P273 - P301 + P312 + P330