Order Entry
Denmark
ContactUsLinkComponent
Pesticide reference standard, Bromfenvinphos-ethyl, PESTANAL®, analytical standard, Supelco®
Pesticide reference standard, Bromfenvinphos-ethyl, PESTANAL®, analytical standard, Supelco®
Catalog # 45816-100MG
Supplier:  Merck
CAS Number:  
undefined
Pesticide reference standard, Bromfenvinphos-ethyl, PESTANAL®, analytical standard, Supelco®
Catalog # 45816-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    33399-00-7
  • Description:
    Bromfenvinphos-ethyl
  • Formula:
    C₁₂H₁₄BrCl₂O₄P
  • Storage temperature:
    2...8 °C
  • Description:
    Bromfenvinphos-ethyl

Specifications

About this item