Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Bis(2-ethylhexyl) isophthalate, analytical standard, Supelco®
Organic reference standard, Bis(2-ethylhexyl) isophthalate, analytical standard, Supelco®
Catalog # 91344-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Bis(2-ethylhexyl) isophthalate, analytical standard, Supelco®
Catalog # 91344-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    137-89-3
  • Description:
    Bis(2-ethylhexyl) isophthalate
  • Formula:
    C₂₄H₃₈O₄
  • Description:
    Bis(2-ethylhexyl) isophthalate

Specifications

About this item