Supelco® Organic Reference Standard, BDE No 47
91834-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:5436-43-1
- Description:BDE No 47
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00626457
- M-Clarity quality level:MQ100
- Synonym(s):2,2′,4,4′-TetraBDE; 2,2′,4,4′-Tetrabromodiphenyl ether; PBDE 47
- Molecular weight:485,79