Organic reference standard, BDE No 203 solution, analytical standard, Supelco®
33687-1ML
: Merck
:
- Concentration:50 µg/ml in isooctane
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:337513-72-1
- Description:BDE No 203 solution
- M-Clarity quality level:MQ100
- Formula:C₁₂H₂Br₈O
- Storage temperature:2...8 °C
- Description:BDE No 203 solution