Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 4-Aminobenzoic acid, analytical standard, Supelco®
Organic reference standard, 4-Aminobenzoic acid, analytical standard, Supelco®
Catalog # 01973-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 4-Aminobenzoic acid, analytical standard, Supelco®
Catalog # 01973-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    150-13-0
  • Description:
    4-Aminobenzoic acid
  • Melting point:
    186...188 °C
  • Formula:
    C₇H₇NO₂
  • Description:
    4-Aminobenzoic acid

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.