Organic reference standard, 1-Nitrosopiperidine solution, certified reference material, Supelco®
Catalog # 40458.
Supplier: Merck
CAS Number:
Specifications
- Concentration:5000 µg/ml in methanol
- Pk:1
- Standards:Certified reference material
- CAS No.:100-75-4
- Description:1-Nitrosopiperidine solution
- Environmentally Preferable:
- Formula:C₅H₁₀N₂O
- Storage temperature:2...8 °C
- Description:1-Nitrosopiperidine solution
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.