Order Entry
Denmark
ContactUsLinkComponent
Poly(Bisphenol A carbonate) analytical standards kit, molecular weight series, for GPC
Poly(Bisphenol A carbonate) analytical standards kit, molecular weight series, for GPC
Catalog # 181676-1SET
Supplier:  Merck
CAS Number:  
undefined
Poly(Bisphenol A carbonate) analytical standards kit, molecular weight series, for GPC
Catalog # 181676-1SET
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    1 SET
  • Description:
    Poly(Bisphenol A carbonate) kit
Specifications
About this item
Poly(Bisphenol A carbonate) (PC) is a thermoplastic material. It is injected in molding operations and is majorly studied to understand its thermal behaviour, especially thermal degradation property.
Kit components:
Average Mw 23000; average Mn 13,000 25 g
Average Mw 31000; average Mn 18,000 25 g
Average Mw 40000; average Mn 21,000 25 g