Order Entry
Denmark
ContactUsLinkComponent
Sodium hexametaphosphate, Macron Fine Chemicals™
Sodium hexametaphosphate, Macron Fine Chemicals™
Catalog # MACRE024-61
CAS Number:  
undefined
Sodium hexametaphosphate, Macron Fine Chemicals™
Catalog # MACRE024-61
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth, black T/E cap
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Phosphorus Pentoxide (P₂O₅):
    ≥65.0 %
  • pH of 1% Solution at 25°C:
    6.7 - 7.2

Specification Test Results

About this item

 :
 :611.77 g/mol
Melting Pt:610 °C
Density:1.25 g/cm³ (20 °C)
EINECS:233-343-1
CAS Number:10124-56-8
Formula:Na₆P₆O₁₈
MDL Number:MFCD00136045