D-(+)-Glucose, white powder, Molecular Biology Grade
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
:180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Optical Rotation (c=10, Water, NH4OH):+52,5 ± 5°
- Moisture:Report Result
Specification Test Results
Frequently Bought Together