Order Entry
Denmark
Orders LinkContactUsLinkComponent
Test Strips, Potassium

Test Strips, Potassium

Potassium iodide starch paper MN 616 T for nitrite and free chlorine, 4 - mg/L - NO₂⁻; 1 - mg/L - Cl₂, Color reaction with potassium iodide/starch, 48×25×84 mm (packaging), 200 Tests, 200
Catalog # MANA90758
Supplier:   MACHEREY-NAGEL
Test Strips, Potassium
Catalog # MANA90758
Supplier:  MACHEREY-NAGEL
CAS Number:  
Test Strips, Potassium
Test Strips, Potassium

Potassium iodide starch paper MN 616 T for nitrite and free chlorine, 4 - mg/L - NO₂⁻; 1 - mg/L - Cl₂, Color reaction with potassium iodide/starch, 48×25×84 mm (packaging), 200 Tests, 200

CAS Number:  
Price
363,00 kr
Log in for availability
1 * 200 Items
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. QUANTOFIX® test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips allow the quick and easy determination of potassium in solutions. They include all necessary reagents so that the measurement can be started immediately. A reliable result is obtained within 1 minute.

Specifications

Specifications

  • Chemical method:
    Color reaction with potassium iodide/starch
  • Pk:
    200
  • Description:
    Potassium iodide starch paper MN 616 T for nitrite and free chlorine
  • Dimensions:
    48×25×84 mm (packaging)
  • Tests per kit:
    200 Tests
  • Range:
    4 - mg/L - NO₂⁻; 1 - mg/L - Cl₂
  • Cat. No.:
    MANA90758
  • Sensitivity:
    Nitrite - 1 mg/L NO₂⁻;Free chorine - 1 mg/L Cl₂
  • Contents:
    200 test strips in a plastic box
  • Platform:
    Qualitative test paper
  • Compliance:
    Not CE certified and no hazardous material
  • Shelf life (months):
    24 (from production)
  • W×D×H:
    20×70 mm (net)
  • Method:
    Nitrite - white → blue-violet;Chlorine - white → blue-violet
  • Weight:
    49,2 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.