Order Entry
Denmark
ContactUsLinkComponent
Test paper, qualitative, Plumbtesmo®
Test paper, qualitative, Plumbtesmo®
Catalog # MANA90602
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test paper, qualitative, Plumbtesmo®
Catalog # MANA90602
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    40
  • Description:
    Plumbtesmo® for lead and lead ions (Pb, Pb²⁺)

Specifications

About this item

The test paper allows the quick and easy detection of lead (Pb) on surfaces including metal surfaces, dishes, ceramics and toys. In criminal investigations it is used to search for traces of bullets.

  • Colour change: White-yellow to pink

The surface in question is moistened with distilled water. The test paper is pressed against the surface for about two minutes. In the presence of Lead the paper turns pink to dark purple.

Limit of sensitivity: 5 mg/L

Packaging: Pack of 40 sheets 40×25 mm