Order Entry
Denmark
ContactUsLinkComponent
Pharmaceutical analytical standard, 2-Thiouracil-13C,15N2, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, 2-Thiouracil-13C,15N2, reference material, Honeywell Fluka™
Catalog # HONCR1508-10MG
CAS Number:  
Pharmaceutical analytical standard, 2-Thiouracil-13C,15N2, reference material, Honeywell Fluka™
Catalog # HONCR1508-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    2-Thiouracil-¹³C,¹⁵N₂
  • Formula:
    C₃¹³CH₄¹⁵N₂OS
  • Storage temperature:
    Ambient
  • Description:
    2-Thiouracil-¹³C,¹⁵N₂
  • Freezing point:
    > 290 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results