Order Entry
Denmark
ContactUsLinkComponent
 
Pharmaceutical analytical standard, 6-Phenyl-2-thiouracil-13C,15N2, reference material, Honeywell Fluka™
  HONCR1501-10MG
 :  
Pharmaceutical analytical standard, 6-Phenyl-2-thiouracil-13C,15N2, reference material, Honeywell Fluka™
  HONCR1501-10MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    6-Phenyl-2-thiouracil-¹³C,¹⁵N₂
  • Formula:
    C₉¹³CH₈¹⁵N₂OS
  • Storage temperature:
    Ambient
  • Description:
    6-Phenyl-2-thiouracil-¹³C,¹⁵N₂
  • Freezing point:
    263...265 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results