Pharmaceutical analytical standard, 5-Nitrobenzimidazole, reference material, Honeywell Fluka™
Catalog # HONCR1395-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- CAS No.:94-52-0
- Description:5-Nitrobenzimidazole
- Formula:C₇H₅N₃O₂
- Storage temperature:Ambient
- Description:5-Nitrobenzimidazole
- Freezing point:204...209 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results