Order Entry
Denmark
ContactUsLinkComponent
Pharmaceutical analytical standard, Albendazol-2-aminosulfone hydrochloride, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Albendazol-2-aminosulfone hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1394-25MG
CAS Number:  
Pharmaceutical analytical standard, Albendazol-2-aminosulfone hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1394-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mg
  • Description:
    Albendazol-2-aminosulfone hydrochloride
  • Formula:
    C₁₀H₁₃N₃O₂S · HCl
  • Storage temperature:
    Ambient
  • Description:
    Albendazol-2-aminosulfone hydrochloride
  • Freezing point:
    241...243 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results