Pesticide reference standard, Fluoroimide, neat, Honeywell Fluka™
HONCR1248-25MG
:
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:41205-21-4
- Description:Fluoroimide
- Formula:C₁₀H₄Cl₂FNO₂
- Storage temperature:Ambient
- Description:Fluoroimide
- Freezing point:240 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results