Order Entry
Denmark
ContactUsLinkComponent
Pharmaceutical analytical standard, Hydroxyfenbendazole monohydrate, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Hydroxyfenbendazole monohydrate, reference material, Honeywell Fluka™
Catalog # HONCR1223-10MG
CAS Number:  
Pharmaceutical analytical standard, Hydroxyfenbendazole monohydrate, reference material, Honeywell Fluka™
Catalog # HONCR1223-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    1584701-11-0
  • Description:
    Hydroxyfenbendazole monohydrate
  • Formula:
    C₁₅H₁₃N₃O₃S·H₂O
  • Storage temperature:
    Ambient
  • Description:
    Hydroxyfenbendazole monohydrate
  • Freezing point:
    215 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results