Order Entry
Denmark
ContactUsLinkComponent
Pharmaceutical analytical standard, Cefpodoxime, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Cefpodoxime, reference material, Honeywell Fluka™
Catalog # HONCR1103-25MG
CAS Number:  
Pharmaceutical analytical standard, Cefpodoxime, reference material, Honeywell Fluka™
Catalog # HONCR1103-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mg
  • CAS No.:
    80210-62-4
  • Description:
    Cefpodoxime
  • Formula:
    C₁₅H₁₇N₅O₆S₂
  • Storage temperature:
    –20 °C
  • Description:
    Cefpodoxime
  • Freezing point:
    200 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results