Safranine O solution for Gram staining, Sigma-Aldrich®
: Merck
:477-73-6
:
:C₂₀H₁₉ClN₄
:MFCD00011759
:350.85 g/mol
:0.98 g/cm3 (20 °C)
:M-Clarity™ quality level = MQ300
:49 °C
:GHS02
:Ambient
:1
- Suitability for microscopy:passes test
- gram-positve microorganisms:blue-violet
- gram-negative microorganisms:pink to red
Frequently Bought Together