Order Entry
Denmark
ContactUsLinkComponent
Chemical pads
Chemical pads
  PRTWSM80YER
 :  Portwest
undefined
Chemical pads
  PRTWSM80YER
 :  Portwest
  • Description:
    Chemical pads, yellow
  • L×W:
    500×400 mm
  • Pk:
    200
Perforated single spun bonded layers gives excellent tensile strength and rapid absorption properties.
  • Suitable for use in lint free environments
  • Each pad can absorb 0,4 L of aggressive and non-aggressive chemicals
 : Meets standards BS 7959-1 and BS 7959-3.