Order Entry
Denmark
ContactUsLinkComponent
Test strips, qualitative, chromium
Test strips, qualitative, chromium
Catalog # MANA90724
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, qualitative, chromium
Catalog # MANA90724
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Diphenylcarbazide
  • Pk:
    200
  • Description:
    Chromium test papers

Specifications

About this item

This test paper allows the quick and easy detection of chromate (CrO₃-). Cr(III) ions must be converted to Chromate prior to the detection.

  • Colour change: White to violet

For non-destructive testing of materials apply a drop of acid solution (1 part hydrochloric acid 37% + four parts hydrogen peroxide 3%) to the degreased surface. After 10 to 30 seconds add a drop sodium hydroxide (NaOH).

Limit of sensitivity: 2 mg/l Cr3+ or 5 mg/l CrO42–

Delivery: Pack of 200 strips 20×70 mm