Propionic acid 99.5-100.5% NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:79-09-4
:
Formula:C₃H₆O₂
MDL Number:MFCD00002756
:74.08 g/mol
Merck Index:13,07917
Boiling Pt:141 °C (1013 hPa)
Melting Pt:–21 °C
Density:0.99 g/cm³ (20 °C)
Flash Pt:49 °C
Storage Temperature:Ambient
EINECS:201-176-3
UN:3463
ADR:8,II
REACH:01-2119486971-24
:GHS02
:GHS05
:GHS06
Specification Test Results
- Assay:Min. 99.5 - 100.5%
- Specific gravity at 25 °C:0.988 - 0.993
- Distilling Range:138.5 - 142.5 °C
- Nonvolatile Residue:0.0001
- Elemental Impurities:As Reported
- Readily Oxidizable Substances:To Pass Test
- Aldehyde:To Pass Test
- Identification:To Pass Test
- Residual Solvents:To Pass Test
Specification Test Results
Frequently Bought Together