2-Methyl-1,3-propanediol ≥98.0% (by GC)
:2163-42-0
:
:C₄H₁₀O₂
:MFCD00082586
:123…125/20 mm
:–54 °C
:1.015 g/cm³
:127 °C (261°F)
:90.12 g/mol
:606-809-0
- Appearance (Colour):Clear colourless to very pale yellow
- Form:Liquid
- Assay (GC):≥98.0%
- Water Content (Karl Fischer Titration):≤0.25%
Frequently Bought Together