Order Entry
Denmark
ContactUsLinkComponent
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (by HPLC) suitable for fluorescence, Sigma-Aldrich®
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (by HPLC) suitable for fluorescence, Sigma-Aldrich®
Catalog # T5428-25MG
CAS Number:  
undefined
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (by HPLC) suitable for fluorescence, Sigma-Aldrich®
Catalog # T5428-25MG
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :500.93 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
CAS Number:115532-50-8
Formula:C₂₅H₂₅N₂O₃·ClO₄
MDL Number:MFCD00274460