Order Entry
Denmark
ContactUsLinkComponent
Wipes, pre-saturated, PROSAT®
Wipes, pre-saturated, PROSAT®
Catalog # 115-2439
Supplier:  CONTEC
undefined
Wipes, pre-saturated, PROSAT®
Catalog # 115-2439
Supplier:  CONTEC
Specifications
  • Packed:
    30 wipes/bag; 36 bags/case
  • Description:
    Meltblown polypropylene LE wipes, presaturated, PROSAT® Sterile™
  • L×W:
    230×280 mm
  • Pk:
    1.080
Specifications
About this item
The melt blown polypropylene wipes provide a consistent release of solvent to thoroughly remove surface contaminants in critical environments.
  • The resealable pouch reduces waste and the pre saturated wipe controls solvent usage and VOCs
  • Suitable for grade A/B or ISO Class 5 environments
  • Melt blown polypropylene fabric is exceptionally clean and free from additives
  • Low in particles and fibres so ideal for product contact areas
  • IPA of about 70% and 30% DI water
Micro sized filaments provide exceptional particle removal and resealable pouch preserves cleanliness and solvent saturation levels.
Certifications: Each lot is tested before release and low endotoxin certified to <20 EU/wipe, eliminating the risk of introducing endotoxins and other contaminants into product contact areas.