
Sodium hydroxide 1.0 M NaOH (1.0N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Assay:0.950 - 1.050 N
- Carbonate (as Na₂CO₃):Max. 0.03 N
- Traceable to NIST SRM:effective
- Vapor Pressure:3 mmHg ( 37 °C)
Specification Test Results
Frequently Bought Together