Order Entry
Denmark
ContactUsLinkComponent
 
Silica gel PS 2-6 mm drying agent, with indicator, Fluka™
Catalog # 78307-1KG
CAS Number:  
Silica gel PS 2-6 mm drying agent, with indicator, Fluka™
Catalog # 78307-1KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Particle Size:
    2-6 mm
  • Appearance:
    complying
  • pH (10 % susp., 20°C):
    3.5 - 7.0
  • Water absorbing capacity:
    Min. 20 %
  • Copper (Cu):
    0.03 - 0.12 %
  • Sieve analysis 2-6 mm:
    Min. 95 %
  • Bulk Density:
    600 - 800 g/l
  • Change:
    complying

Specification Test Results

About this item

 :
Melting Pt:>1.000 °C
Storage Temperature:Ambient
CAS Number:112926-00-8
MDL Number:MFCD00011232