Order Entry
Denmark
ContactUsLinkComponent
Rinsing solution I, CHROMASOLV™ LabReady™, Solvent Blend rinsing agent for LC-MS, Riedel-de Haën™
Rinsing solution I, CHROMASOLV™ LabReady™, Solvent Blend rinsing agent for LC-MS, Riedel-de Haën™
Catalog # 34689-1L
   Safer Human and Environmental Health
CAS Number:  
Rinsing solution I, CHROMASOLV™ LabReady™, Solvent Blend rinsing agent for LC-MS, Riedel-de Haën™
Catalog # 34689-1L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Assay of water:
    49 - 51 Vol.%
  • Identity 2-propanol (GC):
    complying
  • Non-volatile matter:
    Max. 0.001 %
  • Silver (Ag):
    Max. 0.1 ppm
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.1 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Sodium (Na):
    Max. 0.1 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Tin (Sn):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • Organic impurities GC:
    Max. 0.1 Fl.%
  • Absobance at 254 nm:
    Max. 0.008
  • Absorbance at 230 nm:
    Max. 0.045
  • Absorbance at 210 nm:
    Max. 0.096
  • Suitability for the LC-MS:
    complying

Specification Test Results

About this item

 :
Flash Pt:22 °C
Storage Temperature:Ambient
UN:1219
ADR:3,II
 :GHS02
 :GHS07
MDL Number:MFCD00011674