Order Entry
Denmark
ContactUsLinkComponent
 
Potassium hexacyanoferrate(II) trihydrate ≥99%, Purum, Fluka™
Catalog # 12639-25KG
CAS Number:  
Potassium hexacyanoferrate(II) trihydrate ≥99%, Purum, Fluka™
Catalog # 12639-25KG
CAS Number:  

Specifications

  • Pack type:
    Fiberboard box
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Water insoluble matter:
    Max. 0.02 %
  • Chloride (Cl):
    Max. 0.5 %
  • Sulfate (SO₄):
    Max. 0.1 %

Specification Test Results

About this item

 :
 :422.39 g/mol
Melting Pt:70 °C (Elimination of water of crystallisation)
Melting Pt:70 °C (–H₂O of crystall.)
Density:1.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-722-2
CAS Number:14459-95-1
Formula:K₄Fe(CN)₆·3H₂O
MDL Number:MFCD00167023