Order Entry
Denmark
ContactUsLinkComponent
 
Heptane (mixture of isomers) ≥99.8%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Catalog # 01379-1L
CAS Number:  
undefined
Heptane (mixture of isomers) ≥99.8%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Catalog # 01379-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.8 %
  • Water (Karl Fischer):
    Max. 0.005 %
  • Silver (Ag):
    Max. 0.5 ppb
  • Aluminium (Al):
    Max. 1 ppb
  • Arsenic (As):
    Max. 1 ppb
  • Gold (Au):
    Max. 0.5 ppb
  • Barium (Ba):
    Max. 0.5 ppb
  • Beryllium (Be):
    Max. 0.5 ppb
  • Bismuth (Bi):
    Max. 0.5 ppb
  • Calcium (Ca):
    Max. 20 ppb
  • Cadmium (Cd):
    Max. 0.5 ppb
  • Cerium (Ce):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 0.5 ppb
  • Chromium (Cr):
    Max. 1 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 1 ppb
  • Iron (Fe):
    Max. 1 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Germanium (Ge):
    Max. 0.5 ppb
  • Mercury (Hg):
    Max. 1 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Iridium (Ir):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 10 ppb
  • Lithium (Li):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 1 ppb
  • Manganese (Mn):
    Max. 0.5 ppb
  • Molybdenum (Mo):
    Max. 0.5 ppb
  • Sodium (Na):
    Max. 100 ppb
  • Nickel (Ni):
    Max. 1 ppb
  • Lead (Pb):
    Max. 0.5 ppb
  • Palladium (Pd):
    Max. 0.5 ppb
  • Platinum (Pt):
    Max. 0.5 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 0.5 ppb
  • Selenium (Se):
    Max. 1 ppb
  • Tin (Sn):
    Max. 0.5 ppb
  • Strontium (Sr):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 1 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 0.5 ppb
  • Zinc (Zn):
    Max. 1 ppb
  • Zirconium (Zr):
    Max. 0.5 ppb
  • Refractive Index:
    n20/D 1.387 (lit.)

Specification Test Results

About this item

 :
 :100.2 g/mol
Flash Pt:–4 °C
EINECS:205-563-8
UN:1206
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
Boiling Pt:98.2…98.4 °C (1.000 hPa)
Melting Pt:–90.5 °C
Density:0.69 g/cm³ (15 °C)
ADR:3 II
Merck Index:13,04679