Human native collagenase type 3 (from Clostridium histolyticum)
Catalog # ABNOP5274
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 mg
- Conjugation:Unconjugated
- Enzyme type:Native
- Source:Clostridium histolyticum
- Species:Human
- Storage conditions:Store at 4 °C
- Form:Lyophilized powder
- Enzyme name:Collagenase type 3
Specifications
About this item
Collagenases are enzymes that break the peptide bonds in collagen.
- Dialysed
- Recommend usage: collagenase assay, mammary primary cell isolation
Lower in secondary proteolytic contaminant activities but with typical collagenase activity.