
Orthoboric acid ≥99.8% (by acidimetry) ACS, ISO for analysis
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
:61.83 g/mol
EINECS:233-139-2
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay (Acidimetric):Min. 99.8%
- Residue on Ignition (as SO₄) (CH₃OH+HCl):Max. 0.05%
- Heavy Metals (as Pb):Max. 0.0005%
- Insoluble Matter in CH₃OH:Max. 0.005%
- Insoluble matter in H₂O:Max. 0.005%
- Chloride:Max. 0.0005%
- Phosphate:Max. 0.0005%
- Sulphate:Max. 0.002%
- As:Max. 0.00005%
- Ca:Max. 0.002%
- Fe:Max. 0.0001%
- K:Max. 0.005%
- PB:Max. 0.0005%
- Ag:Max. 0.0005%
- Al:Max. 0.0005%
- Cd:Max. 0.0005%
- Co:Max. 0.0005%
- Cr:Max. 0.0005%
- Cu:Max. 0.0005%
- MG:Max. 0.0005%
- Mn:Max. 0.0005%
- Mo:Max. 0.0005%
- Ni:Max. 0.0005%
- Sr:Max. 0.0005%
- Ti:Max. 0.0005%
- Tl:Max. 0.0005%
- V:Max. 0.0005%
- Zn:Max. 0.0005%
- Li:Max. 0.0005%
- Au:Max. 0.0005%
- Be:Max. 0.0005%
- Bi:Max. 0.0005%
- Ga:Max. 0.0005%
- Ge:Max. 0.0005%
- Ba:Max. 0.0005%
- In:Max. 0.0005%
- Pt:Max. 0.0005%
- Sb:Max. 0.0005%
- Si:Max. 0.0005%
- Sn:Max. 0.0005%
- Zr:Max. 0.0005%
Specification Test Results
Frequently Bought Together