Supelco® Organic Reference Standard, CARB Method 1004 DNPH Mix 1
47650-U
: Merck
:
- Concentration:3 µg/ml in acetonitrile (as aldehydes & ketones)
- Pk:1 Ampoul
- Description:CARB Method 1004 DNPH mix 1
- Seq:1
- Storage conditions:2…8 °C
- Hazard class description:H225 - H302 + H312 + H332 - H319
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:45 °C
- GHS signal: GHS02, GHS07
: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338