Order Entry
Denmark
ContactUsLinkComponent
6-CFDA (6-Carboxyfluorescein diacetate, single isomer) ≥99% (by TLC) fluorescent pH indicator
6-CFDA (6-Carboxyfluorescein diacetate, single isomer) ≥99% (by TLC) fluorescent pH indicator
Catalog # 51021.
Supplier:  Biotium
CAS Number:  
undefined
6-CFDA (6-Carboxyfluorescein diacetate, single isomer) ≥99% (by TLC) fluorescent pH indicator
Catalog # 51021.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 6-CFDA, 6-Carboxy-di-O-acetylfluorescein, 6-Carboxyfluorescein diacetate
 :
 :460.39 g/mol
Storage Temperature:Freezer
CAS Number:3348-03-6
Formula:C₂₅H₁₆O₉
MDL Number:MFCD00036871