Order Entry
Denmark
ContactUsLinkComponent
BCECF AM ester 1 mg/mL in anhydrous DMSO ≥95% (by TLC) fluorescent pH indicator
BCECF AM ester 1 mg/mL in anhydrous DMSO ≥95% (by TLC) fluorescent pH indicator
Catalog # 51009.
Supplier:  Biotium
CAS Number:  
undefined
BCECF AM ester 1 mg/mL in anhydrous DMSO ≥95% (by TLC) fluorescent pH indicator
Catalog # 51009.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :808.69 g/mol
Storage Temperature:Freezer
CAS Number:117464-70-7
Formula:C₄₂H₄₀O₂₁
MDL Number:MFCD00036969