Supelco® Organic Reference Standard, 4-(4-Hydroxyphenyl)-2-Butanone
01416-100MG
: Merck
:
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:5471-51-2
- Description:4-(4-Hydroxyphenyl)-2-butanone
- Storage conditions:15...25 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00002394
- M-Clarity quality level:MQ300
- Synonym(s):Frambione; NSC 26515; Raspberry ketone
- Melting point:81...85 °C (lit.)
- Formula:HOC₆H₄CH₂CH₂COCH₃
- Molecular weight:164,20
- Flash point:Not applicable
- GHS signal: GHS07
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P301 + P312 + P330